XCALI-98088 - Xcalibur 4.3 Release Notes
Description:
These release notes briefly list changes in the Xcalibur 4.3 software, fixes to previously reported issues, and known issues still existing in the Xcalibur 4.3 software release.
Environment:
Xcalibur 4.3
Hardware:
- Recommended 3.1 GHz Quad core processor with 16 GB RAM
- Video card and monitor capable of 1920 ×1080 resolution
- 1 TB hard drive
- DVD drive
Software:
- Microsoft Windows 10 Enterprise 2016 LTSB or Windows 7 Professional SP1 64-bit
- .NET Framework 4.6.2
- Thermo Foundation 3.1 SP7 or later
- Microsoft Office 2013
- Adobe Acrobat or Reader 10.1
Attachment(s):
| File | Last Modified |
|---|---|
| XCALI-98088 - Xcalibur 4.3 Release Notes.PDF | December 29, 2021 |