Thermo Fisher Scientific
Shutdown Sequence for Pathfinder EDS Systems
Description: 
Follow these steps to shutdown Pathfinder EDS systems:
  1. Back out the slide using the motor or manual knob
  2. Shutdown Pathfinder software
  3. Power off Peltier Controller (rocker switch by power cable) and confirm LED lights go off
  4. Power off motor controller, if present (rocker switch by power cable)
  5. Power off Pathfinder Analyzer (black button on top)
Environment:
Pathfinder EDS
  • Was this article helpful?