Thermo Fisher Scientific
269-237802 - Rev A - Installing Your Software for RESULT Version 3 Software
Description: 
This manual explains how to install the Thermo Scientific RESULT TQ Analyst, Language Pack and optional ValPro software on a computer you have provided.
Environment:
  • Antaris EX
  • Antaris MX
  • Antaris IGS
  • Antaris II
  • Antaris Target
All of these software applications require both Microsoft Internet Explorer version 6 or higher and Microsoft Windows operating system software with networking services. The applications work with Windows 7 Professional and Windows XP Professional.