Thermo Fisher Scientific
269-238002-5 - Rev A - Workflow Events and Specifications for RESULT Integration Software
Description: 
A functional workflow is comprised of workflow events and their associated specifications. Workflow events specify tasks that will be carried out when the workflow is run. The software carries out the events in the order in which they appear in the workflow navigation frame when you display the workflow in RESULT Integration.
This manual describes the software workflows.
Environment:
  • Antaris EX
  • Antaris MX
  • Antaris IGS
  • Antaris II
  • Antaris Target
All of these software applications require both Microsoft Internet Explorer version 6 or higher and Microsoft Windows operating system software with networking services. The applications work with Windows 7 Professional and Windows XP Professional.