Thermo Fisher Scientific
Diamond Phonon Explanation
Description:
Diamond crystals used during infrared analysis has natural absorbances that occur between 2600 and 1600 cm-1 in the mid-Infrared region and represent the phonon bands within the structure of the diamond crystal.
The attachment describes this phenomena.
Environment:
Nicolet Continuum
  • Was this article helpful?