Thermo Fisher Scientific
Failed to connect to database
Issue
Users not granted access to the database
Environment
  • FT-IR
  • OMNIC
Resolution
The Administrator had to add the group containing the users to the group containing the users to the actual database with connection rights. Refer to the attachment for more details.
  • Was this article helpful?