Thermo Fisher Scientific
Nicolet Summit FTIR Beam Information
Issue
What is the aperture size of the Summit and the diameter of the beam?
Environment
  • Nicolet Summit
  • Nicolet Summit PRO
Resolution
Aperture sizes and beam diameters appear in the table below. Refer to the attachment for more information.
Instrument Model Aperture Aperture Size (mm) IR Beam Diameter Sample Compartment (mm) IR Beam Diameter At Detector (mm)
Summit Fixed Aperture 1.85 5.78 1.16
Summit PRO Large Aperture 2.29 7.15 1.43
Summit PRO Small Aperture 0.81 2.53 0.51