Thermo Fisher Scientific
Silicon Wafer Properties For Use With FTIR
Issue
What are three key properties for the silicon wafer that will give the optimum performance in FTIR analysis?
Environment
FT-IR
Resolution
  • Backside finish should be selected to be low scattering (bright-etch) for best S/N and low variability
  • Resistivity needs to be above 5 ꭥ-cm to maintain optical transparency and eliminate variability from this source
  • Interstitial oxygen should be controlled to minimize spectral variation introduced from the wafer
  • Was this article helpful?