Thermo Fisher Scientific
Replacing a Crystal Plate
Environment:
Smart ATR
Resolution: 
To replace the crystal plate:
  1. Remove the two Phillips or straight blade screws securing the pressure device (circled in red below) and remove the pressure device.
  2. Remove the old plate and install the new plate.
  3. Carefully replace the pressure device and secure with the two screws.
Figure 1 - Replace crystal plate.png
  • Was this article helpful?