Thermo Fisher Scientific
Install-004 - Rev 02 - Installing the iZ10
Description: 
The iZ10 is a second sample compartment and detector that can be added to the right side of the iN10 or iS10. The attached document describes the process for installing the module
Environment:
  • Nicolet iN10 or iS10
  • Nicolet iZ10 FT-IR auxiliary module