Cetac ASX-560, ASX-280 home position error and Labbook fails
Description:
After autosampler ASX-560 or ASX-280 installation, it may occur running a labbook, that Qtegra stops when the probe returns to home position from random rack position, the autosampler led in dashboard turns red, showing false communication issue.
Environment:
- iCAP 6000 Series
- iCAP 7000 Series
- ICAPPRO Series
- Qtegra
- ASX 560
- ASX 280
Attachment(s):
File | Last Modified |
---|---|
Cetac ASX-560_280 home position error Rev 2.pdf | May 09, 2022 |
ASX-280 & ASX-560 Alignment Guide.pdf | May 09, 2022 |