Error 2208
Issue
Error E2208
Environment
- iTEVA
- Qtegra
- ICP-OES
- iCAP 6000 Series
- iCAP 7000 Series
Resolution
The E2208 error is generated when the plasma fails to light but no known interlock has been triggered during the failure to ignite the plasma. When this error is encountered, using the attached guide should help determine what the source of the ignition failure is.
Attachment(s)
File | Last Modified |
---|---|
Troubleshooting Error Code E2208 and E2209.pdf | February 02, 2022 |