Thermo Fisher Scientific
Effects of sample charging
Description: 
A sample is being bombarded with negatively charged electrons and the sample becomes negatively charged because it is not a good conductive path for the electrons in the beam to get to ground through the sample.  
Environment:
  • Noran System 6
  • Noran System 7
  • Pathfinder