Thermo Fisher Scientific
NORAN Light Element X-Ray Detectors
Description: 
High deadtime
Environment:
  • Noran System 6
Resolution:
If the Chamber scope is turned on then turn chamber scope off.
  • Was this article helpful?