Thermo Fisher Scientific
DXR first generation Spectrograph driver
Issue
Drivers from original spectrograph are no longer included in current versions of OMNIC software, which is causing the system to not communicate.
Environment
  • OMNIC versions greater than 9.3
  • Raman DXR
Resolution
Attached is a file with the 64bit drivers from the 9.3 update DVD. Download the attached file and extract files to a folder and update driver from device manager.