Autosampler Rack Error in iTEVA
Description:
After the Win7 update on 11 October 2017, the error “Cannot find object” can occur when creating an autosampler sequence.
Environment:
iTEVA
Attachment(s):
File | Last Modified |
---|---|
Autosampler Rack Error in iTEVA.pdf | December 04, 2021 |