Thermo Fisher Scientific
Moving the stage or turret
The microscope features a motorized stage and turret, which can be moved only by the software or by the optional joystick. Never attempt to move the stage or turret manually.
Avoid pinch hazard.
Keep your hands and fingers away from the motorized turret and the stage area except to position the sample or change accessories. Remove your hands from the system when operating the software.
  • Was this article helpful?