Thermo Fisher Scientific
Connections and ports
raptir rear ports and connections.jpg
A USB 3.0 connection to computer's USB 3.0 port
B Connection to the optional joystick
C Connect to Auxiliary Signals port on the spectrometer
D Connect to "Accessory" port on the spectrometer
E Connection to power supply cable
  • Was this article helpful?