Thermo Fisher Scientific
Create and Customize Microscopy Reports
After analyzing a sample, you can print or save a report to record your findings.
❖ To create a microscopy report
  1. From the Session Analysis tab, go to File > Create Report.
  2. Enter a title for the report, select a format, and choose a template. The available templates depend on the type of analysis you performed.
  3. To customize the report, select Options and choose elements to include or exclude.
  4. Click Create.
  • Was this article helpful?