Create and Share Reports and Export Data
Generate reports from templates to quickly save or share your data as Microsoft™ Word™, PowerPoint™, or Excel™ files, or export your spectra, workflows, or settings.
❖ To create a report from the desktop interface
-
From the Spectra view, select File > Create Report.
-
Enter a title for your report and select a format and template option. The preview updates to match your selections.
If you select Save as your format, your report data is saved to the database.
-
To configure the template, click Options and select items to include. Click OK to save your selection.
-
Click Create.
❖ To create a report from the touchscreen interface
-
From the spectral view, open the menu and select Create Report.
-
Enter a title for your report, select a directory to save the report to, choose a format, and finally select a template. The preview updates to match your format and template selection.
If you select Save as your format, your report data is saved to your database.
-
To configure the template, touch Options and select items to include. Touch OK to save your selection
-
Touch Create.
Export settings, spectra, or workflows
Use OMNIC Paradigm software for desktop to export settings and workflows. Export spectra in either desktop or touchscreen mode.
❖ To export settings
-
Select the settings you wish to export from the Settings list.
-
Select Acquire Data > Export Settings.
-
Name the settings file and click Save.
Settings are saved with a .expx file extension. These settings files store your measurement settings but not Search, QCheck, or Quantification settings. Settings files with a .expx file extension are not compatible with previous versions of OMNIC software.
❖ To export workflows
-
If you are using the desktop interface
a. In the Workflows pane, right-click the workflow you wish to export and select Export from the menu.
b. Name your workflow file and click Save.
c. Workflow files are saved with a .rwfl file extension.
-
If you are using the touchscreen interface
a. In the Workflows tab, select the workflow you wish to export and select the more options menu.
b. Select Export.
c. Name your workflow file and touch Save.
d. Workflow files are saved with a .rwfl file extension.
❖ To export measurements
-
If you are using the desktop interface
a. In the Measurements pane, right-click the spectrum you wish to export and select Export.
b. Name your spectrum file and click Save. Spectra are saved with a .spa file extension.
-
If you are using the touchscreen interface
a. From the home screen, select the Measurements tab.
b. Open the more options menu and select Export.
c. Touch Save. Spectra are saved with a .spa file extension.