Thermo Fisher Scientific
Measure a Background Spectrum
Before you collect sample data, you must measure the background. A background is a measurement of the instrument sampling conditions without any sample present. After measuring the background and the sample, the sample data is ratioed against the background measurement to create the final sample spectrum.
The best location or material to use varies depending on the sample type or application. The table below provides some suggestions for collecting backgrounds.
Sample type and application
Background suggestion
Transmission, sample suspended over air
Air
Transmission, sample on a salt plate
An area of the salt plate where no sample material is present
Transmission, sample in a compression cell
A crystal of KBr (or other halide salt) in the same compression cell next to the sample
Specular reflection
A mirror or polished metal
Diffuse reflection
Crushed KBr, silicon carbide paper or other rough, non-infrared-absorbing material
Reflection-absorption
A mirror or polished metal, or an area of the sample support where no sample material is present
ATR
Air
❖ To measure a background spectrum
  1. Start a session and capture a low-magnification mosaic.
  2. From the floating toolbar, select the Background tool.
  3. Click the mosaic at the point where you want to measure the background. A live single beam spectrum is displayed in the spectra pane. To move the background point, click the mosaic again.
  4. When you are happy with the single beam spectrum of the background, click Accept Background.
  5. Click Measure Background.The background spectrum is collected. When it is complete, it is added to the Background tab of the Spectra tab.
    paradigm-mosaic-background.png
When the background measurement is complete, you are ready to place areas, lines, and points and measure your sample.
If you are measuring multiple areas over a long period of time, replace your background measurement periodically. In general, you should always have a recent measurement of the background before you measure the sample.
  • Was this article helpful?