Thermo Fisher Scientific
View Measurement History
View the measurement history to visually compare a spectrum at different stages of processing and analysis. For example, after applying automatic baseline correction, view the measurement history to quickly see how the spectrum changed.
❖ To view the measurement history
  1. From the Spectral View, navigate to the history panel.
  2. Right-click any item in the history and select Measurement History.
view measurement history.png
The historical data displays the spectrum at each stage in your processing and analysis.
  • To create a report of the historical data, open the File menu and select Create Report. Select the Historical Spectrum template, edit your report settings, and click Create.
  • To exit the measurement history, click Close Measurement History.
  • Was this article helpful?