Thermo Fisher Scientific
AA83863 - Rev A - Auto Sample Changer Instruction Notices
Description: 
All ARL EQUINOX instruments are shipped with a basic non-spinning sample holder suited for powder measurement in reflection mode. This sample holder is necessary for adjustment and alignment and can be used for measurement of samples as well. Attached are instruction notices for the sample changer.
Environment:
  • ARL EQUINOX
  • EQUINOX Sample Changer