Thermo Fisher Scientific
AA83866 - Rev A - RPED Adjustment Bench Instruction Notice
Description: 
ARL EQUINOX X-Ray diffractometers are designed for measuring of crystalline samples (powder, bulk, metals, thin layers, etc...), phase identification, nanoscale structural analysis, non-destructive quality control, as well as for many other applications.
Thermo Fisher Scientific develops and sells many sample holders that can be plugged in their diffractometer to analyze the samples. The sample holders are sometimes with moving parts and some electronics is needed on some more complex devices to control the movement.
The RPED adjustment bench enables users to set two different sample holders: the capillary sample holder and the Gandolfi sample holder. For each sample holder please refer to their instruction notice manuals.
Environment:
ARL EQUINOX