Sample Holder Instruction Notices
Description:
All ARL EQUINOX instruments are shipped with a basic non-spinning sample holder suited for powder measurement in reflection mode. This sample holder is necessary for adjustment and alignment and can be used for measurement of samples as well. Attached are instruction notices for these sample holders.
Environment:
- ARL EQUINOX
- EQUINOX Sample Thin Layer System
- EQUINOX Sample Holders:
- SSRT
- SSRT - Cement
- SSRZ
Attachment(s):
File | Last Modified |
---|---|
AA83871, Sample holder SSCA.pdf | May 16, 2022 |
XR-DOC AA83859-A_EN, [Instruction Notice, Sample Thin Layer System] 1019.pdf | February 09, 2022 |
XR-DOC AA83858-A_EN, [Instruction Notice, Sample Holder SSRZ] 0619.pdf | February 09, 2022 |
XR-DOC AA83856-A_EN, [Instruction Notice, Sample Holder SSRT] 0619.pdf | February 09, 2022 |
XR-DOC AA83857-A_EN, [Instruction Notice, Sample Holder SSRT Cement] 0619.pdf | February 09, 2022 |