Blank Reading High on Furnace
Issue:
Blank Reading Too High
Environment:
- iCE 3400
- iCE 3500
Resolution:
- Clean out wash bottle and refill with new rinse solution.
- Install new autosampler probe (capillary) tip.
- Install new vial reagent cup or vial for blank and make sure it has been acid washed
- Install new piece of capillary tubing: Installing the Furnace Capillary tubing
Cause:
Contamination