Thermo Fisher Scientific
Burner not in place error
Issue
Customer are receiving a Burner not in place error on new systems
Environment
  • iCE 3300
  • iCE 3500
Resolution
Currently new systems have a new design for the burner position optosensor where the sensor is mount from the back vs older systems where the sensor was mount from the front.
Figure 1: Front mounted
The rear mounted sensors are getting damaged if the burner is not carefully installed on the spray chamber.  The front of the sensor is broken off by the burner base on insertion.
Figure 2: Rear mounted sensor damaged
The burner sensor assembly will need to be replaced.
Burner position Optosensor part number: 420217203042
  • Was this article helpful?