Error MD 2018
Issue
Error MD 2018: The spectrometer has failed to set the correct photomultiplier voltage. The optimum output cannot be found.
Environment
- iCE 3300
- iCE 3400
- iCE 3500
Resolution
Switch out lamps.
Analysis
Customer had a Ca/Mg lamp. Had them run a monochromator calibration which passed. When switched back to their Ca only lamp, the error persisted. Had customer use the Ca/Mg lamp and the error was resolved.
Cause
HCL not putting out enough energy.