Thermo Fisher Scientific
Failed to Setup Optics, AA iCE 3000 Series
Issue
Failed to Setup Optics on Antimony 
Environment
  • iCE 3300
  • iCE 3400
  • iCE 3500
Resolution
Performed monochromator calibration using Ca/Mg lamp
Analysis
As and Pb could setup just fine but were using 0.5mm bandpass whereas Sb method was using 0.2mm bandpass. 
Cause
0.2mm slit must have been off which alignment corrected
  • Was this article helpful?