Flame Autosampler not moving
Issue
The Cetac flame autosampler is not moving when controlled by Solaar software. The autosampler is able to be control by the Cetac software through the computer
Environment
- iCE3000 series
- M series
- S series
Resolution
The autosampler should only be connected to the AA system Accessory 1 or Accesory 2. If the autosampler came with an USB cable it should not be connected to the computer at the same time.