Thermo Fisher Scientific
How To Check For ±24V On The EDS2 Board
How To Find & Make A Backup Copy of Noran System Six (NSS), Noran System 7 (NS7) Software License Key
Issue:
Users need to back up the license key for Noran System Six (NSS) or Noran System 7 (NS7) software to ensure it can be easily restored during a software installation.
Environment:
  • Software: Noran System Six (NSS), Noran System 7 (NS7)
  • Required Tools: MS-Word or WordPad
Resolution:
  1. Create a Folder:
    Create a folder called C:\Service if it does not already exist.
  2. Open a Blank Document:
    Open a blank MS-Word (or WordPad) document and leave it open.
  3. Open NSS/NS7 Software:
    Open the NSS or NS7 software and navigate to an existing data project.
  4. Access the License Key:
    Click Help > About Noran System Six (or About Noran System 7).
    clipboard_ed9050f51215fae6bbd8226204e30af6d.png
  5. Copy the License Key:
    Highlight the complete license key in the help dialogue window. Press Ctrl+C to copy the license key.
  6. Paste License Key into Document:
    Navigate to the open MS-Word document. Paste the license key data by pressing Ctrl+V or using Edit > Paste.
  7. Save the Document:
    Save the document in the C:\Service folder and name it NSS License Key.
    Note: Copying the license key text allows it to be easily copied and pasted during software installation.
  8. Optional - Capture Help Window:
    Ensure the help dialogue window is active by clicking
  • Was this article helpful?