MD132- Operation cannot start unless the correct lamp is fitted
Issue
When trying to start a run an error pops up:
MD132- Operation cannot start unless the correct lamp is fitted
Environment
- iCE 3300/ 3400/ 3500
Resolution
This error occurs when there is no lamp in the lamp menu that matches the analyte selected in the method.
- Make sure the correct method for your analysis is loaded
- Check the lamp menu and make sure that either the coded HCL is reading correctly, or if the lamp is uncoded that it has been manually labeled in the correct position in the lamp carousel.
- Run Setup Optics
- Continue to your run
Analysis
Cause