Thermo Fisher Scientific
Parts for AA PM
Issue
Parts Needed to complete PM
Environment
  • iCE 3300
  • iCE 3400
  • iCE 3500
Resolution
Flame PM
  • Seal for end cap - 401317294561
  • Overpressure disc - 942341030111
  • Nebulizer tubing - 942339005421
  • Cr/Ni Standard - 131350647201
  • O-ring kit - 942339005151
  • Was this article helpful?