Add more than 1 sample carousel
When a method is created with over 60 samples, a second carousel will be needed. This article describes how to add and see more than one (1) sample carousel.
Issue
How to add and see more than 1 sample carousel in a method
Environment
- Solaar
- ICE series
- S series
- M series
Resolution
When a method is created with over 60 samples, a second carousel will be needed. To see the loading guide, you will need to select the ASLG (auto sampler loading guide) button. Once displayed, the carousel ID will populate at the top. To change from one carousel to another, use the Previous / Next button to scroll to the one desired.