Automatic Standards Make Up from a Master Standard
Atomic Absorption iCE 3400 or 3500 Series Procedure to have the Furnace Autosampler make up standards automatically from a master standard. Set up the Calibration tab regarding standards, then select the Sampling tab and select ‘Fixed Volume’ for standards prep. Deposit the same volume for each standard, making up the balance with diluent. Click “Volumes” to view what the autosampler will use to create standards. Change calibration, or make changes in the Sampling tab.
Description:
The attached document is a procedure for having the furnace autosampler make up standards automatically from a master standard.
Environment:
- iCE 3400 Series
- iCE 3500 Series
Attachment(s):
| File | Last Modified |
|---|---|
| Automatic Standards Make Up from a Master Standard.pdf | August 02, 2022 |