Deuterium Lamp has failed to light
The links in this article describe how to change the Deuterium lamp. If the Deuterium lamp goes out (on the S-Series or iCE 3300 AA, ICE 3400/3500/M6 MK2), use Deuterium lamp P/N 9423 420 30004.
Issue
Deuterium (D2) lamp has failed to light
Environment
- iCE 3300
- iCE 3400
- iCE 3500
Resolution
Use the relevant guide pertaining to the instrument you have (iCE 3300 or iCE 3400/3500) attached via the links below to replace the Deuterium lamp
Changing the Deuterium Lamp on an S-Series or iCE3300.pdf
Changing the Deuterium Lamp & Alignment ICE 3400_3500.pdf
Attachment(s)
| File | Last Modified |
|---|---|
| Changing the Deuterium Lamp & Alignment ICE 3400_3500.pdf | October 07, 2022 |
| Changing the Deuterium Lamp on an S-Series or iCE3300.pdf | October 07, 2022 |