Thermo Fisher Scientific
Flame Autosampler not moving
This article addresses the issue of the Cetac flame autosampler not responding when controlled by Solaar software, although it functions correctly with the Cetac software on the computer. This issue occurs in the iCE3000, M series, and S series environments. To resolve this, ensure the autosampler is connected exclusively to the AA system's Accessory 1 or Accessory 2 ports. If the autosampler includes a USB cable, it should not be connected to the computer simultaneously.
Issue
The Cetac flame autosampler is not moving when controlled by Solaar software.  The autosampler is able to be control by the Cetac software through the computer
Environment
  • iCE3000 series
  • M series
  • S series
Resolution
The autosampler should only be connected to the AA system Accessory 1 or Accesory 2.  If the autosampler came with an USB cable it should not be connected to the computer at the same time.
  • Was this article helpful?