How To Find and Make A Backup Copy of Noran System Six (NSS), Noran System 7 (NS7) Software License Key
Issue:
Users need to back up the license key for Noran System Six (NSS) or Noran System 7 (NS7) software to ensure it can be easily restored during a software installation.
Environment:
- Software: Noran System Six (NSS), Noran System 7 (NS7)
- Required Tools: MS-Word or WordPad
How to Check for ±24V on the EDS2 Board
Issue
The EDS Power Supply indicates "Red LEDs On" from the Front End Status Display. This suggests a potential issue with the power supply outputs.
Environment
- Product: EDS2 Board
- Instrument: Analyzer Power Supply
- Relevant Outputs: +5V, ±15V, ±24V
- Indication: Front End Status Display showing "Red LEDs On"
Resolution
To troubleshoot the issue with the ±24V power supply on the EDS2 board, follow these steps:
- Use a Digital Voltmeter (DVM):
- Verify the Analyzer Power Supply Outputs +5V, ±15V.
- Confirm that the ±24V are actually present.
- Locate Test Points:
- Identify two crucial test points on the EDS2 board:
- N & P 24V: Regulator outputs
- N & P 24VF: Filtered regulator outputs
- Identify two crucial test points on the EDS2 board:
- Measure Voltage:
- Clip the meter lead onto the terminals indicated in the images provided.
- Ensure accurate voltage readings at these test points.
Images
Analysis (Optional)
No additional analysis provided.
Cause (Optional)
The root cause of the issue is not specified in the provided documents.