How to adjust the impact bead
Loss of sensitivity in the iCE3300, iCE3500, S series, and M6 systems can be caused by a poorly positioned impact bead. To address this issue, follow the attached procedure, which provides detailed instructions on how to adjust the impact bead properly to restore sensitivity.
Issue
Loss of sensitivity can be a result of a poorly positioned impact bead.
Environment
- iCE3300
- iCE3500
- S series
- M6
Resolution
The attached procedure describes how to adjust the impact bead.
Attachment(s)
| File | Last Modified |
|---|---|
| Optimizing the Impact Bead.pdf | August 02, 2022 |