How to align the standard impact bead
In the iCE3300, iCE3500, S Series, and M6 systems, the impact bead must be directly aligned with the nebulizer to effectively aerosolize the sample. To resolve any issues with alignment, follow the attached procedure, which provides detailed instructions on how to properly align the impact bead.
Issue
The impact bead needs to be directly in line with the nebulizer to properly aeresol the sample
Environment
- iCE3300
- iCE3500
- S Series
- M6
Resolution
The attached procedure describes how to properly align the impact bead
Attachment(s)
| File | Last Modified |
|---|---|
| Aligning the standard adjustable impact bead.pdf | August 02, 2022 |