How to change location of standards and samples in iCE 3000 Furnace auto-sampler rack
To customize the positions of standards or samples in the furnace auto-sampler rack for iCE3400, iCE3500, S Series, and M6 systems, use the Solaar software. Open the Method, navigate to the Sequence tab, and click on ASLG (Auto-sampler Loading Guide). From there, you can click on any standard or sample in the rack, drag it, and drop it into the desired position.
Issue
Customize standards or samples positions in Furnace auto-sampler rack.
Environment
- iCE3400
- iCE3500
- S Series
- M6.
Resolution
In the Solaar software, open Method, click on Sequence tab, click on ASLG (Auto-sampler Loading Guide). Click on any standard or sample from rack and drag then drop into any position.
Attachment(s)
| File | Last Modified |
|---|---|
| ASLG.jpg | January 12, 2023 |