How to check sample uptake
Low absorbance in flame can be caused by a blocked nebulizer in the iCE3300, iCE3500, S series, and M6 instruments. To resolve this issue, refer to the attached procedure, which details how to check for proper sample uptake.
Issue
Low absorbance in flame can be cause by by a blocked nebulizer.
Environment
- iCE3300
- iCE3500
- S series
- M6
Resolution
The attached procedure describes how to check for the proper sample uptake.
Attachment(s)
| File | Last Modified |
|---|---|
| Determining Sample Uptake Rate.pdf | August 02, 2022 |