Manually Align burner to optical path
Issue
Low absorbance can occur if the burner is not aligned properly
Environment
- iCE3300
- iCE3500
- S Series
- M6
Resolution
The attached document describes the alignment procedure
Attachment(s)
| File | Last Modified |
|---|---|
| Manual Aligning of the Burner - version 2.pdf | August 02, 2022 |