Perform Monochromator Calibration
Issue
System is failing to setup optics and one of the steps can be a monochromator calibration to correct the issue
Environment
- Solaar
- M series
- S series
- iCE series
Resolution
If a customer owns a Ca/Mg hollow cathode lamp they can perform the calibration themselves, eliminating the need for a service visit. Attached is a procedure to perform the calibration.