Thermo Fisher Scientific
iCAP 6000 Accessory Option - CETAC ASXPRESS PLUS Manuals
Description: 
The ASXPRESS™ increases ICP-AES/ICP-MS analysis throughput by reducing sample delivery/rinse times without affecting measurement time. Multiple sample introduction steps occur simultaneously, resulting in virtual elimination of stabilization and rinse times.
See the attached brochure and manuals for this instrument.
Part Numbers
  • 842347004030  Cetac ASXPRESS – Aqueous Kit for ASX 260/520 Autosamplers
  • 842347004031  Cetac ASXPRESS – Aqueous Kit for EXR-8 Autosampler
  • 842347004032  Cetac ASXPRESS – Oil Kit for ASX-1400 Autosampler
(60/80 rack)
  • 842347004058  Articulated Mounting System
(for iCAP 6000 Series ICP-OES Duo configurations)
Consumables and Spare Parts
842347004033 Carbon Fiber Sample Probe 1.0mm for Aqueous
842347004034 Restrictor Line for Aqueous
842347004035 Aqueous Valve
842347004036 Tee Connector Kit 1/16" barbed connectors (pack of 4)
842347004037 Check valve
842347004038 Fitting Kit (Aqueous and Oil Kits)
842347004039 Stainless Steel Filtered Probe (for Oil Kit)
842347004040 Diaphragm pump (for Oil Kit)
842347004041 Restrictor Line for Oils
842347004042 Oil Valve
842347004046  Pharmed Peripump tubing Kit
842347004047 Sample loop Assembly Aqueous 2mm ID - 0.7ml (Aqueous Kits)
842347004048 Sample loop Assembly Aqueous 2mm ID - 1.0ml (Aqueous Kits)
842347004049 Sample loop Assembly Aqueous 2mm ID - 1.5ml (Aqueous Kits)
842347004050 Sample loop Assembly Aqueous 2mm ID - 2.0ml (Aqueous Kits)
842347004051 Sample loop Assembly Aqueous 2mm ID - 3.0ml (Aqueous Kits)
842347004052 Sample loop Assembly Aqueous 2mm ID - 4.0ml (Aqueous Kits)
842347004053 Sample loop Assembly Aqueous 2mm ID - 5.0ml (Aqueous Kits)
842347004054 Sample loop Assembly Oils 1mm ID - 1.02ml (Oil Kit)
842347004055 Sample loop Assembly Oils 1mm ID - 1.48ml (Oil Kit)
842347004056 Sample loop Assembly Oils 1mm ID - 2.0ml (Oil Kit)
842347004057 Sample loop Assembly Oils 1mm ID - 3.0ml (Oil Kit)
Environment:
  • iCAP 6500 Radial
  • iCAP 6500 Dual
  • iCAP 6300 Radial
  • iCAP 6300 Dual
  • iCAP 6200 Dual
  • Was this article helpful?