Export Methods and Data to a Different iTEVA Database
Issue
How to export data to a different iTEVA database
Environment
- iCAP 6000
- iCAP 7000
- iTeva
Resolution
To export data to a different DB
- Keep the active connection as the original DB where results were first saved.
- Close iTEVA options window.
- Open the Analyst part of iTEVA and open results using the Sample Query option as usual to view previous data. Select Copy to at the bottom of the window once you have selected the required data.
- The following window pops out to select the DB you want the data copied to (for example, the new DB).
- Press OK.
- To copy methods to a DB (Method/Open...), choose a method, and click Copy to.
- The following window pops out to select the DB you want the data copied to (for example, the new DB).
Analysis
Cause
Made a new database and need methods and results from the old database transferred into the new database.
Attachment(s)
File | Last Modified |
---|---|
results screen ss.png | August 02, 2022 |
select db ss.png | August 02, 2022 |
Method copy to.PNG | August 02, 2022 |