Lo P1 Error on Thermoflex 900/1400/2500 Chiller
Issue
Thermoflex 900/1400/2500 chillers displays the "Lo P1" error on the LCD display
Environment
- iCAP 6000
- iCAP 7000
- iCAP Q
- iCAP Qnova
- Thermoflex 900/1400/2500 Chiller
Resolution
- To release the trapped air, use the steps below.
- If releasing trapped air does not resolve the issue, try these actions:
- Replace the water flow sensor
- Confirm that the correct voltage is applied to the Thermoflex chiller (e.g., 115 V or 220 V).
|
|
|
|
|
|
|
Cause
It is possible that air is trapped at the pressure sensor, especially if you notice that water flows into the reservoir before the "Lo P1” error is displayed.