Nebulizer Spray Chamber Connector Kit
Description:
This kit contains the quick connect fitting and attached capillary tubing to connect nebulizer and spray chamber drain to associated peristalic pump tubing.
The attachment Spray chamber Nebulizer connector kit.pdf describes the content of the kit and how it connects to the spray chamber and nebulizer.
P/N 842312051402 Nebulizer/Spray Chamber Connector Kit contains these:
- Adapter PTFE 2 Blk Rings w/integral capillary (chamber drain) 6 EA
- Adapter PTFE 1 Blk Ring w/integral capillary (chamber drain) 6 EA
- Adapter PTFE 1 Red Ring w/integral capillary (neb uptake) 6 EA
- Fitting Neb Gas 4mm inline QC (Ezylok) 3 EA
Environment:
- iCAP 6000 Series
- iCAP 7000 Series
Attachment(s):
File | Last Modified |
---|---|
Spray chamber Nebulizer connector kit.pdf | July 10, 2023 |