Spray Chamber and Nebulizer Connector Kit
Description:
The attachment Spray Chamber and Nebulizer Connector Kit.pdf describes the content of the kit and how it connects to the spray chamber and nebulizer.
Nebulizer/Spray Chamber Connector Kit.pdf (Part Number 842312051402)
Adapter PTFE 1 Blk Ring w/integral capillary (chamber drain, 0.79mm ID x 1.44mOD) - 6 EA
Adapter PTFE 2 Blk Rins w/ integral capillary (chamber drain, 1.27mm ID x 1.88m OD) 6 EA
Adapter PTFE 1 Brn Ring w/integral capillary (neb uptake, 0.5mm ID x 1.11mm OD) 6 EA
Fitting Neb Gas 4mm inline QC (Ezylok) 3 EA
Environment:
- iCAP 6000 Series
- iCAP 7000 Series
Attachment(s):
File | Last Modified |
---|---|
Spray Chamber and Nebulizer Connector Kit.pdf | August 02, 2022 |