iCAP 6000 Accessory Option - CETAC ASXPRESS PLUS Manuals
Description:
The ASXPRESS™ increases ICP-AES/ICP-MS analysis throughput by reducing sample delivery/rinse times without affecting measurement time. Multiple sample introduction steps occur simultaneously, resulting in virtual elimination of stabilization and rinse times.
See the attached brochure and manuals for this instrument.
Part Numbers
- 842347004030 Cetac ASXPRESS – Aqueous Kit for ASX 260/520 Autosamplers
- 842347004031 Cetac ASXPRESS – Aqueous Kit for EXR-8 Autosampler
- 842347004032 Cetac ASXPRESS – Oil Kit for ASX-1400 Autosampler
(60/80 rack)
- 842347004058 Articulated Mounting System
(for iCAP 6000 Series ICP-OES Duo configurations)
Consumables and Spare Parts
| 842347004033 | Carbon Fiber Sample Probe 1.0mm for Aqueous |
| 842347004034 | Restrictor Line for Aqueous |
| 842347004035 | Aqueous Valve |
| 842347004036 | Tee Connector Kit 1/16" barbed connectors (pack of 4) |
| 842347004037 | Check valve |
| 842347004038 | Fitting Kit (Aqueous and Oil Kits) |
| 842347004039 | Stainless Steel Filtered Probe (for Oil Kit) |
| 842347004040 | Diaphragm pump (for Oil Kit) |
| 842347004041 | Restrictor Line for Oils |
| 842347004042 | Oil Valve |
| 842347004046 | Pharmed Peripump tubing Kit |
| 842347004047 | Sample loop Assembly Aqueous 2mm ID - 0.7ml (Aqueous Kits) |
| 842347004048 | Sample loop Assembly Aqueous 2mm ID - 1.0ml (Aqueous Kits) |
| 842347004049 | Sample loop Assembly Aqueous 2mm ID - 1.5ml (Aqueous Kits) |
| 842347004050 | Sample loop Assembly Aqueous 2mm ID - 2.0ml (Aqueous Kits) |
| 842347004051 | Sample loop Assembly Aqueous 2mm ID - 3.0ml (Aqueous Kits) |
| 842347004052 | Sample loop Assembly Aqueous 2mm ID - 4.0ml (Aqueous Kits) |
| 842347004053 | Sample loop Assembly Aqueous 2mm ID - 5.0ml (Aqueous Kits) |
| 842347004054 | Sample loop Assembly Oils 1mm ID - 1.02ml (Oil Kit) |
| 842347004055 | Sample loop Assembly Oils 1mm ID - 1.48ml (Oil Kit) |
| 842347004056 | Sample loop Assembly Oils 1mm ID - 2.0ml (Oil Kit) |
| 842347004057 | Sample loop Assembly Oils 1mm ID - 3.0ml (Oil Kit) |
Environment:
- iCAP 6500 Radial
- iCAP 6500 Dual
- iCAP 6300 Radial
- iCAP 6300 Dual
- iCAP 6200 Dual
Attachment(s):
| File | Last Modified |
|---|---|
| Cetac ASXpress Brochure.pdf | August 02, 2022 |
| 480176 - Rev 1a - ASXpress Plus EXR-8 Installation Guide.pdf | August 02, 2022 |
| 610092 - CETAC ASXpress Plus Quick Installation Guide.pdf | August 02, 2022 |
| 480167 - Rev 6 - CETAC ASXpress Plus Operators Guide.pdf | August 02, 2022 |
| 480166 - Rev 4 - Cetac ASXpress Operator Manual.pdf | August 02, 2022 |